2-(4-Pyridin-4-yl-5-pyrrolidin-2-ylpyrazol-1-yl)ethanol dihydrochloride

Supplier: Matrix Scientific

067589-250MG
101934-296EA 638.46 USD
101934-296
2-(4-Pyridin-4-yl-5-pyrrolidin-2-ylpyrazol-1-yl)ethanol dihydrochloride
2-(4-Pyridin-4-yl-5-pyrrolidin-2-ylpyrazol-1-yl)ethanol dihydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀Cl₂N₄O
MDL Number: MFCD21605888
CAS Number: 1361111-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR