5-Difluoromethyl-3-(4-phenyl-pyrrolidin-3-yl)-1H-pyrazole

Supplier: Matrix Scientific

067587-250MG
101934-292EA 558.46 USD
101934-292
5-Difluoromethyl-3-(4-phenyl-pyrrolidin-3-yl)-1H-pyrazole
5-Difluoromethyl-3-(4-phenyl-pyrrolidin-3-yl)-1H-pyrazole

Hazard: Irritant

Formula: C₁₄H₁₅F₂N₃
MDL Number: MFCD19691459
CAS Number: 1452554-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR