4-[4-Methyl-5-(2-phenoxyethyl)thiazol-2-yl]azepan-4-ol hydrochloride

Supplier: Matrix Scientific

067586-250MG
101934-290EA 558.46 USD
101934-290
4-[4-Methyl-5-(2-phenoxyethyl)thiazol-2-yl]azepan-4-ol hydrochloride
4-[4-Methyl-5-(2-phenoxyethyl)thiazol-2-yl]azepan-4-ol hydrochloride

Hazard: Irritant

Formula: C₁₈H₂₅ClN₂O₂S
MDL Number: MFCD21605886
CAS Number: 1361111-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR