5-(4-Chlorophenyl)-2-pyrrolidin-3-ylmethyl-2H-tetrazole hydrochloride

Supplier: Matrix Scientific

067585-250MG
101934-288EA 476.92 USD
101934-288
5-(4-Chlorophenyl)-2-pyrrolidin-3-ylmethyl-2H-tetrazole hydrochloride
5-(4-Chlorophenyl)-2-pyrrolidin-3-ylmethyl-2H-tetrazole hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅Cl₂N₅
MW: 300.19 g/mol
MDL Number: MFCD16621954
CAS Number: 1229627-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR