(3aR,9aS)-2-Benzyldecahydro-1,4-diazacyclopentacycloocten-5-one

Supplier: Matrix Scientific

067583-250MG
101934-284EA 687.69 USD
101934-284
(3aR,9aS)-2-Benzyldecahydro-1,4-diazacyclopentacycloocten-5-one
(3aR,9aS)-2-Benzyldecahydro-1,4-diazacyclopentacycloocten-5-one

Hazard: Irritant

Formula: C₁₆H₂₂N₂O
MDL Number: MFCD19691653
CAS Number: 1442085-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR