2-[3-(3-Aminomethylphenyl)-4-pyridin-4-ylpyrazol-1-yl]ethanol dihydrochloride

Supplier: Matrix Scientific

067581-250MG
101934-280EA 476.92 USD
101934-280
2-[3-(3-Aminomethylphenyl)-4-pyridin-4-ylpyrazol-1-yl]ethanol dihydrochloride
2-[3-(3-Aminomethylphenyl)-4-pyridin-4-ylpyrazol-1-yl]ethanol dihydrochloride

Hazard: Irritant

Formula: C₁₇H₂₀Cl₂N₄O
MDL Number: MFCD21605884
CAS Number: 1361116-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR