4-Methyl-2-pyrrolidin-3-ylpyrimidine-5-carboxylic acid amide dihydrochloride

Supplier: Matrix Scientific

067574-250MG
101934-266EA 638.46 USD
101934-266
4-Methyl-2-pyrrolidin-3-ylpyrimidine-5-carboxylic acid amide dihydrochloride
4-Methyl-2-pyrrolidin-3-ylpyrimidine-5-carboxylic acid amide dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₆Cl₂N₄O
MW: 279.17 g/mol
MDL Number: MFCD21605879
CAS Number: 1361112-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR