1-(1-Methyl-piperidine-4-carbonyl)azetidine-3-carboxylic acid

Supplier: Matrix Scientific

067573-250MG
101934-264EA 558.46 USD
101934-264
1-(1-Methyl-piperidine-4-carbonyl)azetidine-3-carboxylic acid
1-(1-Methyl-piperidine-4-carbonyl)azetidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₈N₂O₃
MDL Number: MFCD18381550
CAS Number: 1361112-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR