2-(3-Pyrimidin-5-ylbenzyl)pyrrolidine-2-carboxylic acid amide hydrochloride
Supplier: Matrix Scientific
067571-250MG
101934-260EA
558.46
USD
101934-260
2-(3-Pyrimidin-5-ylbenzyl)pyrrolidine-2-carboxylic acid amide hydrochloride
2-(3-Pyrimidin-5-ylbenzyl)pyrrolidine-2-carboxylic acid amide hydrochloride
Hazard: Irritant
Formula:
C₁₆H₁₉ClN₄O |
MDL Number:
MFCD21605877 CAS Number: 1361113-76-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...