1-[2-(4-Fluorophenyl)acetyl]azetidine-3-carboxylic acid(2-oxo-2,3-dihydro-1H-benzoimidazol-5-ylmethyl)amide

Supplier: Matrix Scientific

067570-500MG
101934-258EA 623.08 USD
101934-258
1-[2-(4-Fluorophenyl)acetyl]azetidine-3-carboxylic acid(2-oxo-2,3-dihydro-1H-benzoimidazol-5-ylmethyl)amide
1-[2-(4-Fluorophenyl)acetyl]azetidine-3-carboxylic acid(2-oxo-2,3-dihydro-1H-benzoimidazol-5-ylmethyl)amide

Hazard: Irritant

Formula: C₂₀H₁₉FN₄O₃
MDL Number: MFCD21605876
CAS Number: 1361113-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR