Azetidine-3-carboxylic acid amidetrifluoroacetic acid salt

Supplier: Matrix Scientific

067562-250MG
101934-242EA 476.92 USD
101934-242
Azetidine-3-carboxylic acid amidetrifluoroacetic acid salt
Azetidine-3-carboxylic acid amidetrifluoroacetic acid salt

Hazard: Irritant

Formula: C₆H₉F₃N₂O₃
MDL Number: MFCD21605873
CAS Number: 1361111-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR