Morpholine-2-carboxylic acid cyclopentylamide hydrochloride
Supplier: Matrix Scientific
067560-250MG
101934-238EA
476.92
USD
101934-238
Morpholine-2-carboxylic acid cyclopentylamide hydrochloride
Morpholine-2-carboxylic acid cyclopentylamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₉ClN₂O₂ |
MDL Number:
MFCD16621947 CAS Number: 1229623-59-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...