Morpholine-2-carboxylic acidmethylamide hydrochloride

Supplier: Matrix Scientific

067557-250MG
101934-232EA 476.92 USD
101934-232
Morpholine-2-carboxylic acidmethylamide hydrochloride
Morpholine-2-carboxylic acidmethylamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₃ClN₂O₂
MDL Number: MFCD21605872
CAS Number: 1361112-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR