4-Methyl-2-pyridin-2-yl-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidine dihydrochloride
Supplier: Matrix Scientific
067555-250MG
101934-228EA
561.54
USD
101934-228
4-Methyl-2-pyridin-2-yl-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidine dihydrochloride
4-Methyl-2-pyridin-2-yl-5,6,7,8-tetrahydropyrido[3,4-d]pyrimidine dihydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₆Cl₂N₄ MW: 299.21 g/mol |
MDL Number:
MFCD21605871 CAS Number: 1361116-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...