2-(2'-Methoxybiphenyl-3-ylmethyl)tetrahydrofuran-2-carboxylic acid

Supplier: Matrix Scientific

067554-250MG
101934-226EA 638.46 USD
101934-226
2-(2'-Methoxybiphenyl-3-ylmethyl)tetrahydrofuran-2-carboxylic acid
2-(2'-Methoxybiphenyl-3-ylmethyl)tetrahydrofuran-2-carboxylic acid

Hazard: Irritant

Formula: C₁₉H₂₀O₄
MDL Number: MFCD21605870
CAS Number: 1361113-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR