6-Methyl-1-pyrrolidin-2-ylmethyl-2-trifluoromethyl-1H-quinolin-4-one hydrochloride

Supplier: Matrix Scientific

067546-250MG
101934-210EA 476.92 USD
101934-210
6-Methyl-1-pyrrolidin-2-ylmethyl-2-trifluoromethyl-1H-quinolin-4-one hydrochloride
6-Methyl-1-pyrrolidin-2-ylmethyl-2-trifluoromethyl-1H-quinolin-4-one hydrochloride

Hazard: Irritant

Formula: C₁₆H₁₈ClF₃N₂O
MDL Number: MFCD21605863
CAS Number: 1361115-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR