6a,7,8,9-Tetrahydro-5H-pyrido[3,2-e]pyrrolo[1,2-a]pyrazin-6-one

Supplier: Matrix Scientific

067544-250MG
101934-206EA 476.92 USD
101934-206
6a,7,8,9-Tetrahydro-5H-pyrido[3,2-e]pyrrolo[1,2-a]pyrazin-6-one
6a,7,8,9-Tetrahydro-5H-pyrido[3,2-e]pyrrolo[1,2-a]pyrazin-6-one

Hazard: Irritant

Formula: C₁₀H₁₁N₃O
MDL Number: MFCD04124193
CAS Number: 91622-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR