3-(1-Methyl-1H-imidazol-2-yl)-2,4,5,5a,6,7,8,8a-octahydropyrrolo[3,4-g]indazole trihydrochloride

Supplier: Matrix Scientific

067542-250MG
101934-202EA 638.46 USD
101934-202
3-(1-Methyl-1H-imidazol-2-yl)-2,4,5,5a,6,7,8,8a-octahydropyrrolo[3,4-g]indazole trihydrochloride
3-(1-Methyl-1H-imidazol-2-yl)-2,4,5,5a,6,7,8,8a-octahydropyrrolo[3,4-g]indazole trihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀Cl₃N₅
MDL Number: MFCD21605861
CAS Number: 1361113-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR