Pyrazine-2-carboxylic acid azepan-4-ylamide dihydrochloride

Supplier: Matrix Scientific

067535-250MG
101934-188EA 558.46 USD
101934-188
Pyrazine-2-carboxylic acid azepan-4-ylamide dihydrochloride
Pyrazine-2-carboxylic acid azepan-4-ylamide dihydrochloride

Hazard: Irritant

Formula: C₁₁H₁₈Cl₂N₄O
MDL Number: MFCD21605855
CAS Number: 1361111-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR