2-(2-Aminoethyl)-6-(2,5-dimethylfuran-3-yl)-2H-pyridazin-3-one hydrochloride

Supplier: Matrix Scientific

067529-250MG
101934-176EA 553.85 USD
101934-176
2-(2-Aminoethyl)-6-(2,5-dimethylfuran-3-yl)-2H-pyridazin-3-one hydrochloride
2-(2-Aminoethyl)-6-(2,5-dimethylfuran-3-yl)-2H-pyridazin-3-one hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClN₃O₂
MDL Number: MFCD21605850
CAS Number: 1361115-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR