Dimethyl(4-morpholin-2-yl-[5,5']bipyrimidinyl-2-yl)amine hydrochloride

Supplier: Matrix Scientific

067525-250MG
101934-168EA 476.92 USD
101934-168
Dimethyl(4-morpholin-2-yl-[5,5']bipyrimidinyl-2-yl)amine hydrochloride
Dimethyl(4-morpholin-2-yl-[5,5']bipyrimidinyl-2-yl)amine hydrochloride

Hazard: Irritant

Formula: C₁₄H₁₉ClN₆O
MDL Number: MFCD21605847
CAS Number: 1361113-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR