4-Pyridin-3-ylmethyltetrahydropyran-4-carboxylic acid
Supplier: Matrix Scientific
067510-250MG
101934-138EA
558.46
USD
101934-138
4-Pyridin-3-ylmethyltetrahydropyran-4-carboxylic acid
4-Pyridin-3-ylmethyltetrahydropyran-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₅NO₃ MW: 221.26 g/mol |
MDL Number:
MFCD21605836 CAS Number: 1361111-50-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...