4-[5-(3,4-Difluorophenoxymethyl)-4-methylthiazol-2-yl]azepan-4-ol hydrochloride
Supplier: Matrix Scientific
067503-250MG
101934-124EA
558.46
USD
101934-124
4-[5-(3,4-Difluorophenoxymethyl)-4-methylthiazol-2-yl]azepan-4-ol hydrochloride
4-[5-(3,4-Difluorophenoxymethyl)-4-methylthiazol-2-yl]azepan-4-ol hydrochloride
Hazard: Irritant
Formula:
C₁₇H₂₁ClF₂N₂O₂S |
MDL Number:
MFCD21605831 CAS Number: 1361111-22-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...