[5-(4-Methoxyphenyl)-4-morpholin-2-ylpyrimidin-2-yl]dimethylamine hydrochloride

Supplier: Matrix Scientific

067493-250MG
101934-104EA 476.92 USD
101934-104
[5-(4-Methoxyphenyl)-4-morpholin-2-ylpyrimidin-2-yl]dimethylamine hydrochloride
[5-(4-Methoxyphenyl)-4-morpholin-2-ylpyrimidin-2-yl]dimethylamine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₃ClN₄O₂
MDL Number: MFCD21605823
CAS Number: 1361115-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR