1-((2R,3aS,9aR)-2-Benzyl-decahydro-1,4-diaza-cyclopentacycloocten-1-yl)ethanone

Supplier: Matrix Scientific

067490-250MG
101934-098EA 687.69 USD
101934-098
1-((2R,3aS,9aR)-2-Benzyl-decahydro-1,4-diaza-cyclopentacycloocten-1-yl)ethanone
1-((2R,3aS,9aR)-2-Benzyl-decahydro-1,4-diaza-cyclopentacycloocten-1-yl)ethanone

Hazard: Irritant

Formula: C₁₈H₂₆N₂O
MDL Number: MFCD19691652
CAS Number: 1391536-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR