4-(2-Dimethylamino-4-pyrrolidin-3-ylpyrimidin-5-yl)benzamide hydrochloride

Supplier: Matrix Scientific

067489-250MG
101934-096EA 476.92 USD
101934-096
4-(2-Dimethylamino-4-pyrrolidin-3-ylpyrimidin-5-yl)benzamide hydrochloride
4-(2-Dimethylamino-4-pyrrolidin-3-ylpyrimidin-5-yl)benzamide hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₂ClN₅O
MDL Number: MFCD21605821
CAS Number: 1361113-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR