4-(2-Pyridin-4-ylbenzyl)piperidine-1,4-dicarboxylic acid mono-tert-butyl ester
Supplier: Matrix Scientific
067487-250MG
101934-092EA
558.46
USD
101934-092
4-(2-Pyridin-4-ylbenzyl)piperidine-1,4-dicarboxylic acid mono-tert-butyl ester
4-(2-Pyridin-4-ylbenzyl)piperidine-1,4-dicarboxylic acid mono-tert-butyl ester
Hazard: Irritant
Formula:
C₂₃H₂₈N₂O₄ |
MDL Number:
MFCD21605819 CAS Number: 1361111-45-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...