4-(2-Pyridin-3-ylbenzyl)tetrahydropyran-4-carboxylic acid

Supplier: Matrix Scientific

067485-250MG
101934-088EA 476.92 USD
101934-088
4-(2-Pyridin-3-ylbenzyl)tetrahydropyran-4-carboxylic acid
4-(2-Pyridin-3-ylbenzyl)tetrahydropyran-4-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₉NO₃
MDL Number: MFCD18381458
CAS Number: 1316223-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR