[5-(3-Fluorophenyl)-4-morpholin-2-ylpyrimidin-2-yl]dimethylamine hydrochloride

Supplier: Matrix Scientific

067484-250MG
101934-086EA 476.92 USD
101934-086
[5-(3-Fluorophenyl)-4-morpholin-2-ylpyrimidin-2-yl]dimethylamine hydrochloride
[5-(3-Fluorophenyl)-4-morpholin-2-ylpyrimidin-2-yl]dimethylamine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₀ClFN₄O
MDL Number: MFCD21605818
CAS Number: 1361116-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR