3-[2-(2-Aminopyridin-4-yl)ethyl]piperidine-1-carboxylic acid tert-butyl ester

Supplier: Matrix Scientific

067483-250MG
101934-084EA 687.69 USD
101934-084
3-[2-(2-Aminopyridin-4-yl)ethyl]piperidine-1-carboxylic acid tert-butyl ester
3-[2-(2-Aminopyridin-4-yl)ethyl]piperidine-1-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₇H₂₇N₃O₂
MW: 305.42 g/mol
MDL Number: MFCD21605817
CAS Number: 1361112-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR