2-(2-Amino-6-piperidin-4-ylpyrimidin-4-yl)ethanol dihydrochloride

Supplier: Matrix Scientific

067482-250MG
101934-082EA 476.92 USD
101934-082
2-(2-Amino-6-piperidin-4-ylpyrimidin-4-yl)ethanol dihydrochloride
2-(2-Amino-6-piperidin-4-ylpyrimidin-4-yl)ethanol dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₀Cl₂N₄O
MDL Number: MFCD21605816
CAS Number: 1361112-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR