4-(3-Pyrazin-2-ylpropyl)piperidine-4-carboxylic acid methylamide hydrochloride

Supplier: Matrix Scientific

067480-250MG
101934-078EA 558.46 USD
101934-078
4-(3-Pyrazin-2-ylpropyl)piperidine-4-carboxylic acid methylamide hydrochloride
4-(3-Pyrazin-2-ylpropyl)piperidine-4-carboxylic acid methylamide hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₃ClN₄O
MDL Number: MFCD21605815
CAS Number: 1361115-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR