4-(3-Pyrazin-2-ylpropyl)piperidine-4-carboxylic acid dimethylamide dihydrochloride

Supplier: Matrix Scientific

067474-250MG
101934-066EA 558.46 USD
101934-066
4-(3-Pyrazin-2-ylpropyl)piperidine-4-carboxylic acid dimethylamide dihydrochloride
4-(3-Pyrazin-2-ylpropyl)piperidine-4-carboxylic acid dimethylamide dihydrochloride

Hazard: Irritant

Formula: C₁₅H₂₆Cl₂N₄O
MDL Number: MFCD21605810
CAS Number: 1361116-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR