3-Aminoindan-5-sulfonic acid dimethylamide

Supplier: Matrix Scientific

067467-250MG
101934-052EA 476.92 USD
101934-052
3-Aminoindan-5-sulfonic acid dimethylamide
3-Aminoindan-5-sulfonic acid dimethylamide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂S
MDL Number: MFCD18381514
CAS Number: 1316225-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR