2-Methyl-3-piperidin-3-ylpyrazinetris(trifluoroacetic acid salt)

Supplier: Matrix Scientific

067447-250MG
101934-012EA 638.46 USD
101934-012
2-Methyl-3-piperidin-3-ylpyrazinetris(trifluoroacetic acid salt)
2-Methyl-3-piperidin-3-ylpyrazinetris(trifluoroacetic acid salt)

Hazard: Irritant

Formula: C₁₆H₁₈F₉N₃O₆
MDL Number: MFCD21605796
CAS Number: 1361113-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR