N-Methyl-3-(2-methyl-6-pyrrolidin-2-ylpyridin-4-yl)propionamide

Supplier: Matrix Scientific

067443-250MG
101934-004EA 638.46 USD
101934-004
N-Methyl-3-(2-methyl-6-pyrrolidin-2-ylpyridin-4-yl)propionamide
N-Methyl-3-(2-methyl-6-pyrrolidin-2-ylpyridin-4-yl)propionamide

Hazard: Irritant

Formula: C₁₄H₂₁N₃O
MW: 247.34 g/mol
MDL Number: MFCD21605794
CAS Number: 1361116-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR