1-[4-(2-Chloropyridin-4-ylmethyl)azepan-1-yl]ethanone

Supplier: Matrix Scientific

067439-250MG
101933-996EA 638.46 USD
101933-996
1-[4-(2-Chloropyridin-4-ylmethyl)azepan-1-yl]ethanone
1-[4-(2-Chloropyridin-4-ylmethyl)azepan-1-yl]ethanone

Hazard: Irritant

Formula: C₁₄H₁₉ClN₂O
MDL Number: MFCD19691625
CAS Number: 1316218-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR