1-[3-(5-Bromopyridin-3-ylmethyl)piperidin-1-yl]ethanone

Supplier: Matrix Scientific

067435-250MG
101933-988EA 558.46 USD
101933-988
1-[3-(5-Bromopyridin-3-ylmethyl)piperidin-1-yl]ethanone
1-[3-(5-Bromopyridin-3-ylmethyl)piperidin-1-yl]ethanone

Hazard: Irritant

Formula: C₁₃H₁₇BrN₂O
MDL Number: MFCD19691570
CAS Number: 1316222-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR