4-(4-Acetylpiperazin-2-yl)benzoic acid methyl ester

067433-250MG
101933-984EA 5912.15 USD
101933-984
4-(4-Acetylpiperazin-2-yl)benzoic acid methyl ester
4-(4-Acetylpiperazin-2-yl)benzoic acid methyl ester

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₃
MDL Number: MFCD18381632
CAS Number: 1316226-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR