1-(1-Isopropylpiperidin-4-ylmethyl)-1H-pyrazole-4-carboxylic acid

Supplier: Matrix Scientific

067427-250MG
101933-972EA 558.46 USD
101933-972
1-(1-Isopropylpiperidin-4-ylmethyl)-1H-pyrazole-4-carboxylic acid
1-(1-Isopropylpiperidin-4-ylmethyl)-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₂₁N₃O₂
MDL Number: MFCD18381634
CAS Number: 1316222-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR