4-(4-Carboxypyrazol-1-yl)azepane-1-carboxylic acid tert-butyl ester

Supplier: Matrix Scientific

067426-250MG
101933-970EA 558.46 USD
101933-970
4-(4-Carboxypyrazol-1-yl)azepane-1-carboxylic acid tert-butyl ester
4-(4-Carboxypyrazol-1-yl)azepane-1-carboxylic acid tert-butyl ester

Hazard: Irritant

Formula: C₁₅H₂₃N₃O₄
MW: 309.37 g/mol
MDL Number: MFCD21605791
CAS Number: 1361113-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR