1-[3-(6-Bromopyridin-2-ylmethyl)piperidin-1-yl]ethanone

Supplier: Matrix Scientific

067420-250MG
101933-958EA 558.46 USD
101933-958
1-[3-(6-Bromopyridin-2-ylmethyl)piperidin-1-yl]ethanone
1-[3-(6-Bromopyridin-2-ylmethyl)piperidin-1-yl]ethanone

Hazard: Irritant

Formula: C₁₃H₁₇BrN₂O
MDL Number: MFCD19691507
CAS Number: 1316227-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR