4-(5-Bromo-4-methylthiazol-2-yl)-1-isopropylpiperidine

Supplier: Matrix Scientific

067417-250MG
101933-952EA 476.92 USD
101933-952
4-(5-Bromo-4-methylthiazol-2-yl)-1-isopropylpiperidine
4-(5-Bromo-4-methylthiazol-2-yl)-1-isopropylpiperidine

Hazard: Irritant

Formula: C₁₂H₁₉BrN₂S
MW: 303.27 g/mol
MDL Number: MFCD16621967
CAS Number: 1361116-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR