4-Chloro-6-[2-(1-isopropylpiperidin-2-yl)ethyl]pyrimidine

Supplier: Matrix Scientific

067415-250MG
101933-948EA 558.46 USD
101933-948
4-Chloro-6-[2-(1-isopropylpiperidin-2-yl)ethyl]pyrimidine
4-Chloro-6-[2-(1-isopropylpiperidin-2-yl)ethyl]pyrimidine

Hazard: Irritant

Formula: C₁₄H₂₂ClN₃
MDL Number: MFCD19691504
CAS Number: 1316218-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR