1-[4-(6-Chloropyridin-3-ylmethyl)azepan-1-yl]ethanone

Supplier: Matrix Scientific

067407-250MG
101933-932EA 638.46 USD
101933-932
1-[4-(6-Chloropyridin-3-ylmethyl)azepan-1-yl]ethanone
1-[4-(6-Chloropyridin-3-ylmethyl)azepan-1-yl]ethanone

Hazard: Irritant

Formula: C₁₄H₁₉ClN₂O
MDL Number: MFCD19691623
CAS Number: 1316225-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR