3-(6-Bromopyridin-2-ylmethyl)pyrrolidine-1-sulfonic acid dimethylamide

Supplier: Matrix Scientific

067403-250MG
101933-924EA 638.46 USD
101933-924
3-(6-Bromopyridin-2-ylmethyl)pyrrolidine-1-sulfonic acid dimethylamide
3-(6-Bromopyridin-2-ylmethyl)pyrrolidine-1-sulfonic acid dimethylamide

Hazard: Irritant

Formula: C₁₂H₁₈BrN₃O₂S
MDL Number: MFCD19691649
CAS Number: 1316218-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR