4-Chloro-6-(1-isopropylpiperidin-4-yl)-2-methylpyrimidine

Supplier: Matrix Scientific

067401-250MG
101933-920EA 558.46 USD
101933-920
4-Chloro-6-(1-isopropylpiperidin-4-yl)-2-methylpyrimidine
4-Chloro-6-(1-isopropylpiperidin-4-yl)-2-methylpyrimidine

Hazard: Irritant

Formula: C₁₃H₂₀ClN₃
MDL Number: MFCD19691501
CAS Number: 1316221-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR