4-Chloro-6-[2-(1-isopropylpyrrolidin-2-yl)ethyl]pyrimidine

Supplier: Matrix Scientific

067399-250MG
101933-916EA 558.46 USD
101933-916
4-Chloro-6-[2-(1-isopropylpyrrolidin-2-yl)ethyl]pyrimidine
4-Chloro-6-[2-(1-isopropylpyrrolidin-2-yl)ethyl]pyrimidine

Hazard: Irritant

Formula: C₁₃H₂₀ClN₃
MDL Number: MFCD19691503
CAS Number: 1316225-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR