4-(5-Bromopyrazin-2-ylmethyl)-1-isopropylazepane

Supplier: Matrix Scientific

067389-250MG
101933-896EA 558.46 USD
101933-896
4-(5-Bromopyrazin-2-ylmethyl)-1-isopropylazepane
4-(5-Bromopyrazin-2-ylmethyl)-1-isopropylazepane

Hazard: Irritant

Formula: C₁₄H₂₂BrN₃
MW: 312.26 g/mol
MDL Number: MFCD19691498
CAS Number: 1316218-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR