2-Chloro-4-[2-(1-isopropylpiperidin-2-yl)ethyl]pyridine

Supplier: Matrix Scientific

067387-250MG
101933-892EA 558.46 USD
101933-892
2-Chloro-4-[2-(1-isopropylpiperidin-2-yl)ethyl]pyridine
2-Chloro-4-[2-(1-isopropylpiperidin-2-yl)ethyl]pyridine

Hazard: Irritant

Formula: C₁₅H₂₃ClN₂
MDL Number: MFCD19691494
CAS Number: 1316217-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR